ESD safety ankle boots, lace-up, Business, Nikolas XW Mid
Supplier: ELTEN
Certificates
About this item
Wide, very comfortable business ESD safety ankle boot made from cowhide leather plus a mono-density PU sole (Business).
- Light leather lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (Business ESD)
- Protective steel toecap (200 J)
- Puncture resistant sole (S3 model)
- Extra wide version (XW)
Ideal for use in areas where exposure to moisture is expected.
Colour: Blue, brown
Certifications: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC or S3.
Specifications
- Catalog No:
- ELTN763371/47
- ELTN763371/42
- ELTN763371/48
- ELTN763371/44
- ELTN763371/46
- ELTN763371/41
- ELTN763371/43
- ELTN763371/40
- ELTN763371/45
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001