


Methylmagnesium Iodide 3 M in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:917-64-6
:
Formula:CH₃IMg
MDL Number:MFCD00001026
:166.24 g/mol
Density:1.26 g/cm³
Flash Pt:–40 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to grey to brown Clear to hazy solution
- Strength:2.85 to 3.50 M
- Refractive index:1.4920 to 1.4940 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Related Information
Frequently Bought Together