
2-Methyl-3-phenyl-2-propen-1-ol 95%
:1504-55-8
:
:C₁₀H₁₂O
:MFCD00004738
:148.21 g/mol
:77 °C (.1 mmHg)
:1.03 g/cm³
:113 °C
:GHS07
:Ambient
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5690 to 1.5710 (20°C, 589 nm)
- GC:≥94 %
Frequently Bought Together