


Tribromoboranemethyl sulphide 1 M in methylene chloride, AcroSeal®
Catalog # 428321000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Strength:0.95 to 1.10 M
- Melting point:185 to 189 °C
- HPLC:≥97.5 %
Specification Test Results
About this item
CAS Number:29957-59-3
:
Formula:C₂H₆BBr₃S
MDL Number:MFCD00043296
:312.652 g/mol
Density:1.456 g/cm³
UN:2922
ADR:8,II
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient