


Xylol (Isomerengemisch), extra trocken 98+%, AcroSeal®
:1330-20-7
:
:C₆H₄(CH₃)₂
:MFCD00077264
:106.17 g/mol
:140 °C (1013 hPa)
:–34 °C
:0.865 g/cm³
:23 °C
:1307
:3,III
:GHS02
:GHS07
:GHS08
:Raumtemperatur
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.0 %
- Residue after evaporation:≤0.002 %
Frequently Bought Together