Order Entry
Switzerland
ContactUsLinkComponent
Pyridine for HPLC
Irritant
Flammable
Pyridine for HPLC
Catalog # ACRO396800010
CAS Number:  
undefined
Irritant
Flammable
Pyridine for HPLC
Catalog # ACRO396800010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.8 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.001 %

Specification Test Results

About this item

 :
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.978 g/cm³
Flash Pt:17 °C
Storage Temperature:Ambient
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS07
 :GHS02
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732