
Sarcosine methyl ester hydrochloride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13515-93-0
:
Formula:C₄H₉NO₂·HCl
MDL Number:MFCD00038876
Melting Pt:110…125 °C
:139.58 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤1.0 % (1 g, 70°C) (to constant weight)
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together