ContactUsLinkComponent
(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
Flammable
Corrosive
Irritant
(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
Certificates
undefined
Flammable
Corrosive
Irritant
(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.002 %
  • GC:
    ≥99.0 %
  • Stabilizer:
    150 to 400 ppm BHT
  • Peroxides:
    ≤50 ppm