(±)-1-Phenyl-2-propyn-1-ol 98+%
: 1-Phenyl-2-propyn-1-ol
:4187-87-5
:
:C₉H₈O
:MFCD00021860
:132.16 g/mol
:22 °C
:99 °C
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5450 to 1.5530 (20°C, 589 nm)
- GC:≥98.0 %
Frequently Bought Together