

4-Pentyn-2-ol 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2117-11-5
:
Formula:C₅H₈O
MDL Number:MFCD00004555
:84.12 g/mol
Boiling Pt:126 °C (1013 hPa)
Density:0.896 g/cm³
Flash Pt:37 °C
Storage Temperature:Ambient
UN:1986
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4380 (20°C, 589 nm)
- GC:≥97.0 %
Specification Test Results
Frequently Bought Together