
Tetrabutylammonium chloride 95% tech. contains also bromide
:1112-67-0
:
:C₁₆H₃₆ClN
:MFCD00011635
:277.92 g/mol
:83…86 °C
:1,05 g/cm³ (20 °C)
:110 °C
:GHS07
:Ambient
- Appearance:White to brown Crystalline solid
- Infrared spectrum:Conforms
- Titration Argentometric:≥94 % (on dry substance)
- Water:≤4 %
- Bromide (Br):≤5 %
Frequently Bought Together