ContactUsLinkComponent
Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Certificates
undefined
Silica gel C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Particle size:
    40 to 63 µm
  • Elemental Analysis:
    ≥11.6 % (C)
  • Loading:
    ≥1.21 mmol/g
  • Volatile matter:
    ≤4 % (110°C)