
Bis(2,4-pentanedionato)molybdenum(VI) dioxide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17524-05-9
:
Formula:C₁₀H₁₄MoO₆
MDL Number:MFCD00011506
:326.18 g/mol
Melting Pt:184 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to grey-green to light brown Crystalline powder
- Infrared spectrum:Conforms
- Elemental Analysis:≥96.0 %
Specification Test Results
Frequently Bought Together