Order Entry
Switzerland
ContactUsLinkComponent
Ethyl cellulose, 10 cps 48% (ethoxyl content) 10 cps
Ethyl cellulose, 10 cps 48% (ethoxyl content) 10 cps
Certificates
undefined
Ethyl cellulose, 10 cps 48% (ethoxyl content) 10 cps

Specification Test Results

  • Appearance:
    White to yellow Powder or granules
  • Infrared spectrum:
    Conforms
  • Assay:
    47.5 to 49.5 % (Ethoxyl content)
  • Viscosity:
    8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)

Specification Test Results

Related Information

Frequently Bought Together