ContactUsLinkComponent
Sulphuric acid-[D₂] 98% (w/w) in D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Corrosive
Sulphuric acid-[D₂] 98% (w/w) in D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Certificates
undefined
Corrosive
Sulphuric acid-[D₂] 98% (w/w) in D₂O for NMR spectroscopy (D₂O 99.5+ atom%D)
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • D-Enrichment:
    ≥99.50 %
  • Titration with NaOH:
    97.5 to 102.5 %
  • GC:
    ≥94.0 %