Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Quercetin hydrate 95%

Quercetin hydrate 95%

Quercetin hydrate 95%
Certificates
Quercetin hydrate 95%

3 Options Available Below

Product Details & Documents

Appearance: Yellow Crystalline powder

Synonyms:  3,3',4',5,7-Pentahydroxyflavone hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one xhydrate
  • CAS Number:849061-97-8
  • Formula:C₁₅H₁₀O₇·xH₂O
  • MDL Number:MFCD00006828
  • Molecular Weight:302.24 g/mol (anhy)
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤12 % (K.F.)
  • HPLC:
    ≥94 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.