


Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2802-68-8
:373-57-9
Formula:BF₃·2CH₃OH
MDL Number:MFCD00071635
:131.89 g/mol
Boiling Pt:59 °C (4 mmHg)
Melting Pt:–20 °C
Density:1.222 g/cm³
Flash Pt:68 °C
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Assay:50 to 53 wt% (BF3)
- Density:(20°C) 1.210 to 1.235 kg/l
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤100 ppm
- Arsenic (As):≤1 ppm
- Iron (Fe):≤30 ppm
- Sodium (Na):≤0.1 %
- Heavy metals (as Pb):≤10 ppm
- Nitrogen compounds:≤0.001 %
Specification Test Results
Frequently Bought Together