
Silver phosphate 98%
Catalog # ACRO419370250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Yellow to yellow-green to dark yellow Powder
- Assay:≥97.5 %
- Particle size:≤5.0 % (> 1180 micron)
- Harmonic mean size:0.600 to 0.750 mm
- Uniformity coefficient:≤1.60
- Moisture holding capacity:49.0 to 60.0 % (Cl)
- Total volume capacity:≥1.20 eq/l (Cl)
Specification Test Results
About this item
:
:418.58 g/mol
Melting Pt:849 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7784-09-0
Formula:Ag₃O₄P
MDL Number:MFCD00014148