Order Entry
Switzerland
ContactUsLinkComponent
Dextran for biochemistry, low fraction
Dextran for biochemistry, low fraction
Certificates
undefined
Dextran for biochemistry, low fraction

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤7.0 %
  • UV:
    (6 % in water)
  • Viscosity:
    0.22 to 0.27 (1 % at 37°C)
  • Ash:
    ≤2.0 %
  • Average molecular weight:
    60000 to 90000
  • pH:
    4.5 to 7.0 (6 % in water)

Specification Test Results

Related Information

Frequently Bought Together