Order Entry
Switzerland
ContactUsLinkComponent
 
Toxic
Health
Flammable
Potassium methanolate 0.1 N in toluene/methanol, pure
Catalog # ACRO240605000
CAS Number:  
undefined
Toxic
Health
Flammable
Potassium methanolate 0.1 N in toluene/methanol, pure
Catalog # ACRO240605000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Titration with HCl:
    0.095 to 0.105 N

Specification Test Results

About this item

 :
 :70.13 g/mol
Density:0.85 g/cm³
Flash Pt:7 °C
Storage Temperature:Ambient
UN:1992
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
CAS Number:865-33-8
Formula:CH₃KO
MDL Number:MFCD00012416