Order Entry
Switzerland
ContactUsLinkComponent
Ethyl cellulose, 300 cps 48% (ethoxyl content)
Ethyl cellulose, 300 cps 48% (ethoxyl content)
Certificates
undefined
Ethyl cellulose, 300 cps 48% (ethoxyl content)

Specification Test Results

  • Appearance:
    White Granular powder
  • Infrared spectrum:
    Conforms
  • Assay:
    48 to 49.5 % (ethoxyl)
  • Viscosity:
    250 to 350 cP (5 % in 80/20 toluene/ethanol at 25°C)

Specification Test Results

Related Information

Frequently Bought Together