
L-Methionine sulphoximine 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15985-39-4
:
Formula:C₅H₁₂N₂O₃S
MDL Number:MFCD00002621
:180.222 g/mol
Melting Pt:210 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Specific optical rotation:+12° to +15° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:98.0 to 102.0 %
Specification Test Results
Frequently Bought Together