Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Certificates
About this item
System for Analytical Quality Assurance (AQA), consisting of one combined standard/spiking solution and two round robin test solutions.
Using the standard solution you can easily verify your analytical approach including sampling, sample preparation, pipetting and photometric analysis. The two additional solutions contain unknown concentrations of the analyte and are designed to be used for a participation in a free round-robin test.
Delivery: Each Addista kit comes with one standard solution and two solutions with an unknown concentration to be used with Hach’s LCK cuvette tests.
Frequently Bought Together