ContactUsLinkComponent
Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
  734-2144
undefined
Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
  734-2144
  • Description:
    Vent/close caps for 175 cm² flasks
  • Pk:
    100
Ideal for cell culture flasks. Offered in two different types vent/close and filter.
  • Sterile
  • HDPE closure