ContactUsLinkComponent
Reagent case for soil analysis, VISOCOLOR®
Reagent case for soil analysis, VISOCOLOR®
Catalog # MANA934220
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Reagent case for soil analysis, VISOCOLOR®
Catalog # MANA934220
Supplier:  MACHEREY-NAGEL
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    VISOCOLOR® reagent case for soil analysis with PF-3
Specifications
About this item
This portable laboratory case contains all reagents, instruments and accessories required for the preparation of soil extracts and the subsequent analysis of phosphate (P), potassium (K), ammonium, nitrate, nitrite (N), soil structure and pH.
  • Designed for rapid, convenient and reliable soil analysis in the lab or in the field
  • Measurements are carried out either with colorimetric rapid tests or with easy to use test strips
Ordering information: NANOCOLOR® reagents and photometers must be ordered separately.