Order Entry
Switzerland
ContactUsLinkComponent
Recombinant protein A/G, peroxidase conjugated, Pierce™
Recombinant protein A/G, peroxidase conjugated, Pierce™
Catalog # PIER32490
CAS Number:  
Recombinant protein A/G, peroxidase conjugated, Pierce™
Catalog # PIER32490
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Description:
    Recombinant Protein A/G, peroxidase conjugated
  • Seq:
    05

Specifications

About this item

Pierce™ Recombinant Peroxidase Conjugated Protein A/G is useful to probe and detect antibodies, especially IgG isotypes, of many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognised by either Protein A or Protein G

Gene fusion of the Fc-binding domains of Protein A and Protein G has resulted in production of a structurally and functionally chimeric protein with broader binding than either Protein A or Protein G alone. During fusion, the Protein G gene sequence coding for the serum albumin-binding site is eliminated. This protein is expressed in E. coli and secreted into the surrounding medium during fermentation. The product obtained is consistent in quality and yield because the bacterial host is engineered to be deficient in major proteolytic activities.