Fluorescent dye removal columns, Pierce™
Katalognr. PIER22858
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10
- Description:Fluorescent dye removal columns
Specifications
About this item
Pierce™ Dye Removal Columns enable fast and efficient removal of non-reacted fluorescent dyes from protein labeling reactions.
- Removes free, unconjugated dye from labeled protein solutions in less than a minute
- Purification resin provides outstanding conjugate recovery
- Separate resin and microcentrifuge columns allow optimisation for different dyes and concentrations of sample
Removing excess dye after a labeling reaction is often difficult and time-consuming but is essential for accurate determination of dye-to-protein ratios. The dye removal resin in this kit is highly specialised to produce exceptional protein recoveries while effectively removing non-conjugated dye. Using the appropriate amount of resin and buffer conditions, almost any fluorescent dye can be removed with this kit.