Auftragserfassung
Switzerland
ContactUsLinkComponent
Coupling resin, CarboxyLink™
Coupling resin, CarboxyLink™
Katalognr. PIER20266
CAS-Nummer:  
undefined
Coupling resin, CarboxyLink™
Katalognr. PIER20266
CAS-Nummer:  
Technische Daten
  • VE:
    25 ml
  • Bezeichnung:
    CarboxyLink™ coupling resin
  • Environmentally Preferable:
Technische Daten
Über diesen Artikel
CarboxyLink™ coupling resin is used for covalent immobilisation of peptides or other carboxyl-containing molecules to a porous, beaded resin for use in affinity purification procedures.
  • DADPA-activated crosslinked 4% beaded agarose
  • Couple 1 to 2 mg of peptide per ml of resin
  • Immobilises peptides with high capacity and various orientations without steric hindrance
  • Immoblisation results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin