
Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Storage Temperature:Ambient
EINECS:0
UN:1824
ADR:8,II
:GHS05
Frequently Bought Together