Potassium acetate in aqueous solution USP Test Solution (TS) 34
Catalog # 85364.260
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Acetic acid potassium salt
:
:98.14 g/mol
Merck Index:13,07687
Melting Pt:304 °C
Storage Temperature:Ambient
EINECS:0
CAS Number:127-08-2
Formula:CH₃COOK
MDL Number:MFCD00012458