Order Entry
Switzerland
ContactUsLinkComponent
Phosphotungstic acid 1% USP Test Solution (TS)
Phosphotungstic acid 1% USP Test Solution (TS)
Catalog # 85363.260
Supplier:  VWR Chemicals
CAS Number:  
undefined
Phosphotungstic acid 1% USP Test Solution (TS)
Catalog # 85363.260
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :2880.05 g/mol
Storage Temperature:Ambient
EINECS:0
ADR:8,III
CAS Number:1343-93-7
Formula:H₃O₄₀PW₁₂