ContactUsLinkComponent
ESI negative ion calibration solution, LTQ, Pierce™
ESI negative ion calibration solution, LTQ, Pierce™
Catalog # PIER88324
CAS Number:  
undefined
ESI negative ion calibration solution, LTQ, Pierce™
Catalog # PIER88324
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Description:
    Pierce™ negative ion calibration solution, LTQ ESI Pierce™
  • Seq:
    03
Specifications
About this item
Ion calibration solution are mixtures of highly purified ionizable molecules specifically designed for negative mode calibration of ion trap and orbitrap instruments.
  • Mass spectrometry grade reagents in a non-leachable container
  • Load the mass reference standard into a syringe and inject into the instrument
  • Strong peaks—mixture of SDS, sodium taurocholate and Ultramark 1621 in a acetonitrile/methanol/acetic solution.
It is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific mass spectrometers.