ContactUsLinkComponent
Immobilised TCEP disulphide reducing gel, Pierce™
Immobilised TCEP disulphide reducing gel, Pierce™
Catalog # PIER77712
CAS Number:  
undefined
Immobilised TCEP disulphide reducing gel, Pierce™
Catalog # PIER77712
CAS Number:  
Specifications
  • Pk:
    5 ml
  • Description:
    Immobilised TCEP disulfide reducing gel
Specifications
About this item
Immobilised TCEP disulphide Reducing Gel eliminates the need to use laborious and inefficient gel filtration methods to separate the reduced sample from the reducing agent.
  • Odourless and virtually no loss of activity when stored refrigerated
  • Reduces peptides and proteins over a broad range of pH, salt or detergent concentrations, and temperature conditions
  • Resin slurry provides flexibility for use in spin tube, column or batch modes
  • No soluble reductant removal saves time and conserves sample
  • Cleaves both natural disulfide bonds and those incorporated by crosslinking