ContactUsLinkComponent
Immobilised reductant column
Immobilised reductant column
Catalog # PIER77701
CAS Number:  
undefined
Immobilised reductant column
Catalog # PIER77701
CAS Number:  
Specifications
  • Pk:
    2 ml
  • Description:
    Immobilised reductant column
Specifications
About this item
Pierce™ Immobilised reductant column contains beaded resin to which an thiol-based reducing agent has been immobilised to enable solid-phase reduction of peptide and protein disulphide bonds.
  • Eliminates soluble components which interfere with the assay of free thiol groups
  • No need to remove reducing agent after the reaction
  • Reduced protein, free of contaminating thiols, can be easily assayed using the BCA method
  • Solid phase; protocols for both peptides and proteins
  • Excellent protein recovery
  • Regenerable matrix useable up to four times