ContactUsLinkComponent
Enhanced chemifluorescent HRP substrate kit, QuantaRed™
Enhanced chemifluorescent HRP substrate kit, QuantaRed™
Catalog # PIER15159
CAS Number:  
undefined
Enhanced chemifluorescent HRP substrate kit, QuantaRed™
Catalog # PIER15159
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    QuantaRed™ Enhanced chemifluorescent HRP substrate kit, sufficient for 960 wells (10×96-well plates) at 100 µl working volume
Specifications
About this item
QuantaRed™ Enhanced Chemifluorescent HRP Substrate contains chemical enhancers to increase the fluorescent yield and sensitivity of 10-acetyl-3,7-dihydroxyphenoxazine (ADHP) chemifluorescence.
  • Low picogram to femtogram detection
  • Minimal autofluorescence interference from biological samples
  • Use Stop Solution to effectively stop the reaction resulting in stable signal for at least 4 hours
  • Produces a strong colorimetric signal and can be read fluorescently or colorimetrically at 570 nm
ADHP is a non-fluorescent compound that reacts with horseradish peroxidase (HRP) to produce resorufin, a soluble, highly fluorescent reaction product with excitation/emission maxima of ~570/585nm. The long wavelength emission of the resorufin minimises interference from the low wavelength (blue and green) autofluorescence, making it an ideal substrate for quantitating target molecules in biological samples.
Delivery information: Kit includes QuantaRed™ ADHP Concentrate (1 ml), QuantaRed™ Enhancer Solution (50 ml), QuantaRed™ Stable Peroxide Solution (50 ml), QuantaRed™ Stop Solution (10 ml).