Horseradish Peroxidase, Biotinylated, Pierce™
Catalog # PIER29139
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 mg
- Description:Horseradish peroxidase, biotinylated
Specifications
About this item
Biotinylated horse radish peroxidase is used as control or signal amplification in IHC to amplify the signal of biotinylated primary antibodies using the avidin-biotin complex staining method.