Affinity chromatography media, HisPur™ Ni-NTA Agarose
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
HisPur™ Ni-NTA resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
- High-quality, stable and resilient affinity support
- Bind up to 60 mg of 6xHis-tagged protein per ml of resin
- Purify proteins using native or denaturing conditions
- Same batch of resin can be reused at least five times
HisPur™ Ni-NTA resin consists of beaded agarose derivatised with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni2+). The immobilised metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
Frequently Bought Together