Test kits, chlorine dioxide, Spectroquant®, Supelco®
Catalog # 1.00608.0001
Supplier: Merck
CAS Number:
Specifications
- Chemical method:DPD
- Pk:200 Tests
- Description:Chlorine dioxide test
- M-Clarity quality level:MQ100
- For unit:Prove, Nova 60, Multy, Move, Picco
- Range:0,020 - 10,00 mg/l ClO₂
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.