Orders LinkContactUsLinkComponent
Carcass sampling kit | Avantor
Carcass sampling kit
Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl), 200
Katalognr. TSCLTS/15-BNACL
Anbieter:   TSC LIMITED
Carcass sampling kit
Katalognr. TSCLTS/15-BNACL
Anbieter:  TSC LIMITED
CAS Nummer:  
Carcass sampling kit
Carcass sampling kit
Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl), 200
CAS Nummer:  
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 200 ST
Product Details & Documents
Carcass hygiene blue sampling sponge in easy open stomacher pouch dosed with 0.9% saline.
  • Blue cellulose sponge
  • Easy open stomacher pouch
  • Sterilised by gamma irradiation at dosage of 25-53.4 kGy
Recommended for:
Meat Carcass sampling when testing for Salmonella spp. Conforming to EC Commision 2073/2005 on foodstuffs & compliant with ISO 17604:2003
Zertifizierung: EC 2073/2005 on foodstuffs and compliant with ISO 17604:2003.
Verpackung: Individually packed & dosed with 0.9% Sodium Chloride (NaCl). 200 per box.
Achtung! Bitte beachten: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Technische Daten
Technische Daten
  • Pk:
    200
  • Description:
    Sterile blue sponge in easy open stomacher pouch, individually packed & dosed with 0.9% Sodium Chloride (NaCl)
Technische Daten