Orders LinkContactUsLinkComponent
Detergent solutions, Surfact-Amps™
Detergent solutions, Surfact-Amps™
Detergent solutions, Surfact-Amps™
Certificates
Detergent solutions, Surfact-Amps™
Detergent solutions, Surfact-Amps™
18 Options Available Below
Product Details & Documents
Surfact-Amps™ detergents are ready-to-use, diluted solutions of purified detergent that can be used in routine and high-demand protein research methods and molecular biology techniques. These detergents are useful as a component of cell lysis buffers or other solutions intended to extract and solubilise proteins, improve chromatography, prepare yeast spheroplasts, and reduce background in ELISA or Western blot methods.
  • Clear and pure 10% detergent solutions with <1.0 μeq/ml peroxides and carbonyl content
  • Ampoules are packed under nitrogen for maximum shelf life and are pre-scored for easy opening
  • Easy to pipette and accurately dispense
  • Packaging eliminates the accumulation of peroxides and degradation products
Detergents are a class of molecules whose unique properties enable manipulation (disruption or formation) of hydrophobichydrophilic interactions among molecules in biological samples. In biological research, detergents are used to lyse cells (release soluble proteins), solubilise membrane proteins and lipids, control protein crystallisation and prevent non specific binding in affinity purification and immunoassay procedures. They are often used as an additive in protein gel electrophoresis.
Documents
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • PIER85111
    Triton X-100 Surfact-Amps™ detergent solution
    Triton X-100 Surfact-Amps™ detergent solution
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 281.00
    SFr. 281.00
    1 * 50 mL
       SFr. 281.00
  • PIER85112
    Triton X-100 Surfact-Amps™ detergent solution
    Triton X-100 Surfact-Amps™ detergent solution
    250 ml
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - SFr. 838.00
    SFr. 838.00
    1 * 250 mL
       SFr. 838.00
  • PIER28313
    Triton™ X-100 Surfact-Amps™ detergent solution
    Triton™ X-100 Surfact-Amps™ detergent solution
    1.000 ml
    Log in for availability
    Log in for availability
    1 * 1.000 mL
    1 * 1.000 mL - SFr. 3’010.00
    SFr. 3’010.00
    1 * 1.000 mL
       SFr. 3’010.00
  • PIER28332
    Triton™ X-114 Surfact-Amps™ detergent solution, 6×10 ml
    Triton™ X-114 Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 605.00
    SFr. 605.00
    1 * 1 SET
       SFr. 605.00
  • PIER85113
    Tween™ 20 Surfact-Amps™ detergent solution
    Tween™ 20 Surfact-Amps™ detergent solution
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 285.00
    SFr. 285.00
    1 * 50 mL
       SFr. 285.00
  • PIER85114
    Tween™ 20 Surfact-Amps™ detergent solution
    Tween™ 20 Surfact-Amps™ detergent solution
    250 ml
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - SFr. 1’020.00
    SFr. 1’020.00
    1 * 250 mL
       SFr. 1’020.00
  • PIER85115
    Tween™ 20 Surfact-Amps™ detergent solution
    Tween™ 20 Surfact-Amps™ detergent solution
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 1’650.00
    SFr. 1’650.00
    1 * 500 mL
       SFr. 1’650.00
  • PIER28321
    Tween™ 20 Surfact-Amps™ detergent solution
    Tween™ 20 Surfact-Amps™ detergent solution
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 3’470.00
    SFr. 3’470.00
    1 * 1 L
       SFr. 3’470.00
  • PIER28320
    Tween™ 20 Surfact-Amps™ detergent solution, 6×10 ml
    Tween™ 20 Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 331.00
    SFr. 331.00
    1 * 1 SET
       SFr. 331.00
  • PIER28329
    Tween™ 80 Surfact-Amps™ detergent solution
    Tween™ 80 Surfact-Amps™ detergent solution
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 295.00
    SFr. 295.00
    1 * 50 mL
       SFr. 295.00
  • PIER28230
    Tween™ 80 Surfact-Amps™ detergent solution
    Tween™ 80 Surfact-Amps™ detergent solution
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 1’750.00
    SFr. 1’750.00
    1 * 500 mL
       SFr. 1’750.00
  • PIER28328
    Tween™ 80 Surfact-Amps™ detergent solution, 6×10 ml
    Tween™ 80 Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 326.00
    SFr. 326.00
    1 * 1 SET
       SFr. 326.00
  • PIER85117
    Brij™-35 Surfact-Amps™ detergent solution
    Brij™-35 Surfact-Amps™ detergent solution
    50 ml
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 293.00
    SFr. 293.00
    1 * 50 mL
       SFr. 293.00
  • PIER85118
    Brij™-35 Surfact-Amps™ detergent solution
    Brij™-35 Surfact-Amps™ detergent solution
    500 ml
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 1’570.00
    SFr. 1’570.00
    1 * 500 mL
       SFr. 1’570.00
  • PIER28316
    Brij™-35 Surfact-Amps™ detergent solution, 6×10 ml
    Brij™-35 Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 331.00
    SFr. 331.00
    1 * 1 SET
       SFr. 331.00
  • PIER28336
    Brij™-58 Surfact-Amps™ detergent solution, 6×10 ml
    Brij™-58 Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 418.00
    SFr. 418.00
    1 * 1 SET
       SFr. 418.00
  • PIER85125
    NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution
    NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 1’570.00
    SFr. 1’570.00
    1 * 500 mL
       SFr. 1’570.00
  • PIER28324
    NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution, 6×10 ml
    NP-40 (Nonidet P-40) Surfact-Amps™ detergent solution, 6×10 ml
    1 SET
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - SFr. 326.00
    SFr. 326.00
    1 * 1 SET
       SFr. 326.00