ContactUsLinkComponent
Freund's complete adjuvant, Imject™
Freund's complete adjuvant, Imject™
Catalog # PIER77140
CAS Number:  
undefined
Freund's complete adjuvant, Imject™
Catalog # PIER77140
CAS Number:  
Specifications
  • Pk:
    1 SET
  • Description:
    Imject™ Freund's complete adjuvant, 5×10 ml
Specifications
About this item
Imject™ Freund's Complete Adjuvant (FCA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Water-in-oil emulsion containing killed Mycobacterium
Delivery information: Supplied ready to use in 10 ml ampoules.