ContactUsLinkComponent
Control agarose resin, Pierce™
Catalog # PIER26150
CAS Number:  
undefined
Control agarose resin, Pierce™
Catalog # PIER26150
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Control agarose resin
Specifications
About this item
Pierce™ Control Agarose Resin is underivatised beaded agarose support for pre-clearing protein immunoprecipitation samples.
  • Reduce background signal and non specific binding in IP and Co-IP experiments
  • Resin is supplied as a 50% slurry in 20% ethanol
  • Crosslinked 4% beaded agarose (CL4B) is the same base support as used in Pierce™ IP and co-IP kits
The resin is effective for depleting proteins and other sample components that bind to agarose non specifically, which reduces background signal in IP reactions. When used as a negative control, these beads enable specific and nonspecific binding interactions to be distinguished.