ContactUsLinkComponent
Visual colour kits, chlorine dioxide, MQuant®, Supelco®
Visual colour kits, chlorine dioxide, MQuant®, Supelco®
  1.18754.0001
 :  Merck
 :  
undefined
Visual colour kits, chlorine dioxide, MQuant®, Supelco®
  1.18754.0001
 :  Merck
 :  
  • Chemical method:
    DPD
  • Pk:
    300 Tests
  • Description:
    Chlorine dioxide test
  • M-Clarity quality level:
    MQ100
  • Type:
    MCC
  • Range:
    0,020-0,050-0,075-0,10-0,15-0,20-0,30-0,40-0,55 mg/l ClO₂
Quick tests, visual colour comparison.
  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
 : Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.