SATA (N-Succinimidyl S-Acetylthioglycolate), Pierce™
Catalog # PIER26102
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:231.22 g/mol
CAS Number:76931-93-6
Formula:C₈H₉NO₅S
MDL Number:MFCD00036891