Order Entry
Switzerland
ContactUsLinkComponent
Bovine casein hydrolysate acid, MP Biomedicals
Bovine casein hydrolysate acid, MP Biomedicals
Catalog # ICNA0219102301
Supplier:  MP Biomedicals
CAS Number:  
Bovine casein hydrolysate acid, MP Biomedicals
Catalog # ICNA0219102301
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    0,45 kg
  • Species:
    Bovine
  • Seq:
    01
  • Storage conditions:
    Store at Room Temperature (15 - 30 °C).
  • Form:
    Off-White Powder
  • Protein synonyms:
    Caseamino acids|Hydrochloric acid hydrolysate|casein acid hydrolysate|Amicase
  • Protein/peptide name:
    Casein Hydrolysate Acid

Specifications

About this item

Soluble in water(25 mg/ml).

Product is prepared by acid digestion of casein.

Nitrogen : ~13,0% ; Amino Nitrogen : ~9,2%

Casein acid hydrolysate is used as a nutrient mixture in peptone agars for a variety of bacteria, including Bacillus subtilis, Enterobacter aerogenes, Escherichia coli, Saccharomyces cerevisiae, Staphylococcus aureus, Streptomyces pyogenes, and Streptomyces griseus. It used as a microbiological nutrient in laboratory media and fermentations. It has also been used as a nutrient in vaccine and toxin production where a high free amino acid content is required.