Specifications
- Pk:200 AU
- CAS No.:9029-44-1
- Species:Cucumis sp.
- Seq:01
- Storage conditions:0 °C
- Form:Light blue lyophilized powder
- Enzyme name:Ascorbate oxidase
- Enzyme activity:≥200 unit/mg solid
- Molecular weight:~140 kDa
- pH:5,6
Specifications
About this item
Ascorbate Oxidase is an enzyme belonging to the family of oxidoreductases that catalyses the reaction: Ascorbic acid +¹/₂ O₂ ascorbate oxidase → Dehydroascorbic acid +H₂O.
- Enzyme Commission Number: E.C. 1.10.3.3
- Isoelectric point (pI): between 6,0 and 7,8, 8,2 (Lit.)
Substrate specificity: The enzyme oxidises ascorbic acid and several ascorbic acid derivatives.
It is useful for enzymatic determination of ascorbic acid and for eliminating the interference of ascorbic acid in clinical analysis.
Unit definition: One unit will oxidise 1,0 µmole of L-ascorbate to dehydroascorbate per minute at pH 5,6 and 20 °C.
Stabilizers: Bovine serum albumin (BSA), borax, basic amino acids
Michaelis Constant: 2,5 × 10⁻⁴ M (Ascorbate)
Inhibitors: Azide, cyanide, Na₂S, diethyldithiocarbamate (Na)
Optimum temperature: ~30 °C
Thermal stability: Below 40 °C (pH 8,0, 30 min)