β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) ≥98%
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Nicotinamide adenine dinucleotide reduced, disodium salt (reduced form), NADH disodium salt, ((2R,3S,4R,5R)-5-(6-Aminopurin-9-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy-((((2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxy-oxolan-2-yl)methoxy)hydroxyphosphoryl)oxyphosphinic acid disodium salt
CAS Number:606-68-8
:
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200
:709.41 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Synonyms:β-NADH, 2Na; Reduced DPN; Cozymase, reduced form; Dihydrophosphopyridine nucleotide; Dihydronicotinamide adenine dinucleotide; Diphosphopyridine nucleotide, reduced form; β-DPNH; DPNH; NADH
- Form:White Powder
- Storage:Store at −0 °C. Store desiccated. Protect from light. Store under nitrogen.
- pH:7,5 - 9,5 (10% aq soln)
Specification Test Results
Frequently Bought Together