Order Entry
Switzerland
ContactUsLinkComponent
Collagenase, MP Biomedicals
Catalog # ICNA0215070501
Anbieter:  MP Biomedicals
CAS-Nummer:  
Collagenase, MP Biomedicals
Catalog # ICNA0215070501
Anbieter:  MP Biomedicals
CAS-Nummer:  

Technische Daten

  • Pk:
    1 g
  • CAS No.:
    9001-12-1
  • Form:
    Powder
  • Enzyme name:
    Collagenase
  • Grade:
    Cell Preparation Grade

Technische Daten

Über diesen Artikel

Collagenase can be solubilised in calcium-free solutions such as Hank's Balanced Salts or Earles Balanced Salt Solution.

Unit definition: One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5.