Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Caffeic acid

Fiber drum, 25 kg
Catalog # SPCMCA112-25KGBL
Supplier:   Spectrum Chemical
Caffeic acid
Catalog # SPCMCA112-25KGBL
Supplier:  Spectrum Chemical
CAS Number:  331-39-5
Caffeic acid

Fiber drum, 25 kg

CAS Number:  331-39-5
Price
Inquire For Price
Log in for availability
1 * 25 kg
Product Details & Documents

BSE/TSE statement (available upon request) Allergen statement (available upon request)

Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Merck Index:12,01673
  • Boiling Pt:366 °C (1013 hPa)
  • Melting Pt:234…237 °C
  • EINECS:206-361-2
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Fiber drum
  • Pk:
    25 kg

Specifications