Chromium(III) chloride hexahydrate ≥98% USP
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Chromium trichloride hexahydrate, Chromic chloride hexahydrate
CAS Number:10060-12-5
:
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660
:266.45 g/mol
Merck Index:13,02242
Melting Pt:85 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-038-3
UN:3260
ADR:8,III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98% Min.
- INSOLUBLE MATTER:max. 0.01%
- IRON (Fe):max. 0.01%
- SULFATES (SO4):max. 0.01%
Specification Test Results
Frequently Bought Together